Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erlin2
Entrez Gene ID
244373
Gene Name
ER lipid raft associated 2
Gene Aliases
BC036333, C87251, Spfh2
Chromosome Location
Chr.8: 27023266 - 27039437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153592.2 NP_705820.1 5 - 6 419 104
GenBank mRNA AK030557.1 - 5 - 6 363 104
BC036333.1 - 5 - 6 384 104
BC038374.1 - 5 - 6 359 104

View Full Product Details