Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc9c
Entrez Gene ID
70387
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
2210019E14Rik, 6330408J23Rik
Chromosome Location
Chr.19: 8809066 - 8819780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027412.3 NP_081688.2 2 - 3 524 85
GenBank mRNA AK018145.1 - 2 - 3 564 85
AK169288.1 - 2 - 3 525 85
AK169296.1 - 2 - 3 525 85
BC035304.1 - 1 - 2 435 85
BC043061.1 - 1 - 2 836 85
BC058967.1 - 1 - 2 836 85

View Full Product Details