The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023684.2 | NP_076173.1 | 1 - 2 | 1082 | 71 |
GenBank mRNA | AF115339.1 | - | 1 - 2 | 1082 | 71 |
AK012881.1 | - | 1 - 2 | 146 | 71 | |
BC023065.1 | - | 1 - 2 | 426 | 71 |