Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a9
Entrez Gene ID
228993
Gene Name
solute carrier family 17, member 9
Gene Aliases
1700019H03Rik, BC019537, Vnut
Chromosome Location
Chr.2: 180725213 - 180742280 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183161.3 NP_898984.3 9 - 10 1003 71
GenBank mRNA AK006122.1 - 4 - 5 368 71
BE850231.1 - 2 - 3 132 71

View Full Product Details