Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc17a9
Entrez Gene ID
228993
Gene Name
solute carrier family 17, member 9
Gene Aliases
1700019H03Rik, BC019537, Vnut
Chromosome Location
Chr.2: 180725213 - 180742280 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183161.3 NP_898984.3 10 - 11 1121 59
GenBank mRNA AK006122.1 - 5 - 6 486 59
BE850231.1 - 3 - 4 250 59

View Full Product Details