The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011667.2 | NP_035797.1 | 3 - 4 | 273 | 103 |
GenBank mRNA | AF150963.1 | - | 3 - 4 | 273 | 103 |
AK030031.1 | - | 3 - 4 | 272 | 103 |