Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ifnar2
- Entrez Gene ID
-
15976
- Gene Name
- interferon (alpha and beta) receptor 2
- Gene Aliases
-
AI747302, Ifnar-2
- Chromosome Location
-
Chr.16: 91372215 - 91405589 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
Y09864.1 |
- |
7 - 8 |
924 |
80 |
View Full Product Details