The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025584.2 | NP_001020755.1 | 4 - 5 | 1768 | 73 |
GenBank mRNA | AK081109.1 | - | 4 - 5 | 1503 | 73 |
U11859.1 | - | 4 - 5 | 1727 | 73 | |
U51122.1 | - | 4 - 5 | 1720 | 73 |