Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc9c
Entrez Gene ID
70387
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
2210019E14Rik, 6330408J23Rik
Chromosome Location
Chr.19: 8809066 - 8819780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027412.3 NP_081688.2 1 - 2 276 77
GenBank mRNA AK169288.1 - 1 - 2 277 77
AK169296.1 - 1 - 2 277 77

View Full Product Details