The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011402.3 | NP_035532.2 | 12 - 13 | 1511 | 80 |
GenBank mRNA | AF081499.1 | - | 12 - 13 | 1508 | 80 |
AK004832.1 | - | 12 - 13 | 1510 | 80 | |
AK145599.1 | - | 12 - 13 | 1510 | 80 | |
BC096369.1 | - | 11 - 12 | 1482 | 80 |