The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145965.1 | NP_001139437.1 | 2 - 3 | 321 | 81 |
NM_172910.3 | NP_766498.2 | 3 - 4 | 420 | 81 | |
GenBank mRNA | AK032564.1 | - | 3 - 4 | 421 | 81 |
BB622956.1 | - | - | 421 | 81 | |
BC118969.2 | - | 2 - 3 | 107 | 81 |