Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf32
Entrez Gene ID
56874
Gene Name
ring finger protein 32
Gene Aliases
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2
Chromosome Location
Chr.5: 29195170 - 29225526 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289757.1 NP_001276686.1 2 - 3 188 117
NM_021470.6 NP_067445.3 2 - 3 160 117
NR_110371.1 2 - 3 188 117
GenBank mRNA AK005795.1 - 2 - 3 115 117
AK012288.1 - 2 - 3 188 117
AK016026.1 - 2 - 3 131 117
BC050964.1 - 2 - 3 103 117
BC061103.1 - 2 - 3 126 117
BC089493.1 - 2 - 3 103 117
BY293373.1 - 2 - 3 159 117

View Full Product Details