Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srp72
Entrez Gene ID
66661
Gene Name
signal recognition particle 72
Gene Aliases
5730576P14Rik, 72kDa, AI132477, BC019196, C77589
Chromosome Location
Chr.5: 76974701 - 76999935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025691.1 NP_079967.1 8 - 9 848 100
GenBank mRNA AK133784.1 - 8 - 9 849 100
AK164641.1 - 2 - 3 206 100
BC003434.1 - 8 - 9 836 100
BC019196.1 - 8 - 9 841 100

View Full Product Details