The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201413.1 | NP_001188342.1 | 4 - 5 | 587 | 59 |
NM_001201414.1 | NP_001188343.1 | 4 - 5 | 650 | 59 | |
NM_001201415.1 | NP_001188344.1 | 4 - 5 | 587 | 59 | |
NM_009686.2 | NP_033816.1 | 4 - 5 | 587 | 59 | |
GenBank mRNA | AK004792.1 | - | 4 - 5 | 441 | 59 |
AK032770.1 | - | 4 - 5 | 453 | 59 | |
AK153260.1 | - | 4 - 5 | 616 | 59 | |
AK167649.1 | - | 4 - 5 | 452 | 59 | |
AK169495.1 | - | 4 - 5 | 440 | 59 | |
AK169959.1 | - | 4 - 5 | 528 | 59 | |
BC076587.1 | - | 4 - 5 | 433 | 59 |