The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177561.3 | NP_808229.1 | 6 - 7 | 843 | 71 |
GenBank mRNA | AK029604.1 | - | 5 - 6 | 695 | 71 |
AK145298.1 | - | 6 - 7 | 863 | 71 | |
BC030888.1 | - | 4 - 5 | 610 | 71 | |
BC039916.1 | - | 6 - 7 | 845 | 71 |