Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpl21
Entrez Gene ID
19933
Gene Name
ribosomal protein L21
Gene Aliases
8430440E03Rik, L21
Chromosome Location
Chr.5: 146832890 - 146837032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK168998.1 - 1 - 2 376 101

View Full Product Details