Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm47
Entrez Gene ID
245945
Gene Name
RNA binding motif protein 47
Gene Aliases
9530077J19Rik
Chromosome Location
Chr.5: 66016549 - 66151954 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC034195.1 - 1 - 2 47 64

View Full Product Details