Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem55a
Entrez Gene ID
72519
Gene Name
transmembrane protein 55A
Gene Aliases
2610319K07Rik, AI315591, AV001360
Chromosome Location
Chr.4: 14864219 - 14915260 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028264.4 NP_082540.1 6 - 7 747 90
GenBank mRNA AK012054.1 - 6 - 7 707 90
AK033845.1 - 6 - 7 710 90
AK049213.1 - 6 - 7 748 90
AX775959.1 - 6 - 7 676 90
BC021435.1 - 6 - 7 692 90

View Full Product Details