Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map2k7
Entrez Gene ID
26400
Gene Name
mitogen-activated protein kinase kinase 7
Gene Aliases
5930412N11Rik, JNKK 2, Jnkk2, MAPKK 7, MEK 7, Mapkk7, Mek7, Mkk7, Prkmk7, sek2
Chromosome Location
Chr.8: 4238740 - 4247897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291778.1 NP_001278707.1 1 - 2 189 102
NM_001291783.1 NP_001278712.1 1 - 2 189 102
GenBank mRNA AF060943.1 - 1 - 2 189 102
U93030.1 - 1 - 2 189 102

View Full Product Details