The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183315.2 | NP_899138.1 | 1 - 2 | 222 | 54 |
GenBank mRNA | AK154120.1 | - | 1 - 2 | 173 | 54 |
AK158759.1 | - | 1 - 2 | 116 | 54 | |
BC028881.1 | - | 1 - 2 | 158 | 54 | |
DN176231.1 | - | 1 - 2 | 222 | 54 |