Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc10a2
Entrez Gene ID
20494
Gene Name
solute carrier family 10, member 2
Gene Aliases
ASBT, ISBT
Chromosome Location
Chr.8: 5085623 - 5105232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011388.2 NP_035518.1 2 - 3 544 73
GenBank mRNA AB002693.1 - 2 - 3 544 73
AY529655.1 - 2 - 3 544 73
BC120703.1 - 2 - 3 525 73
BC125290.1 - 2 - 3 525 73
D87059.1 - 2 - 3 471 73

View Full Product Details