Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Letm1
Entrez Gene ID
56384
Gene Name
leucine zipper-EF-hand containing transmembrane protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 33739675 - 33782704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019694.1 NP_062668.1 1 - 2 235 122
GenBank mRNA AB041582.1 - 1 - 2 248 122
AF061026.1 - 1 - 2 235 122
AK133849.1 - 1 - 2 254 122
AK172441.1 - - 270 122
BC023862.1 - 1 - 2 232 122
BC046326.1 - 1 - 2 242 122
BC055865.1 - 1 - 2 253 122
BC061115.1 - 1 - 2 248 122
BC061690.1 - 1 - 2 235 122

View Full Product Details