Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Letm1
Entrez Gene ID
56384
Gene Name
leucine zipper-EF-hand containing transmembrane protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 33739675 - 33782704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019694.1 NP_062668.1 9 - 10 1619 77
GenBank mRNA AB041582.1 - 9 - 10 1632 77
AF061026.1 - 9 - 10 1619 77
AK133849.1 - 9 - 10 1639 77
AK161638.1 - 3 - 4 1341 77
BC023862.1 - 9 - 10 1616 77
BC046326.1 - 9 - 10 1626 77
BC055865.1 - 9 - 10 1637 77
BC061115.1 - 9 - 10 1632 77
BC061690.1 - 9 - 10 1619 77

View Full Product Details