Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3bp2
Entrez Gene ID
24055
Gene Name
SH3-domain binding protein 2
Gene Aliases
3BP2
Chromosome Location
Chr.5: 34525784 - 34563639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145858.1 NP_001139330.1 8 - 9 1459 72
NM_001145859.1 NP_001139331.1 8 - 9 1462 72
NM_011893.3 NP_036023.3 8 - 9 1359 72
GenBank mRNA AK089455.1 - 8 - 9 1290 72
AK143548.1 - 8 - 9 1464 72
AK150265.1 - 8 - 9 1461 72
AK155103.1 - 8 - 9 1316 72
AK155898.1 - 8 - 9 1359 72
AK155970.1 - 8 - 9 1407 72
BC010198.1 - 8 - 9 1297 72
BC085497.1 - 8 - 9 1350 72
L14543.1 - 8 - 9 1293 72

View Full Product Details