Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poln
Entrez Gene ID
272158
Gene Name
DNA polymerase N
Gene Aliases
POL4P
Chromosome Location
Chr.5: 34007179 - 34169526 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK135493.1 - 1 - 2 135 69

View Full Product Details