Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif24
Entrez Gene ID
109242
Gene Name
kinesin family member 24
Gene Aliases
4933425J19Rik, 9430029L23Rik
Chromosome Location
Chr.4: 41390748 - 41464933 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016916.1 - 2 - 3 192 105

View Full Product Details