The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015767.3 | NP_056582.1 | 4 - 5 | 673 | 105 |
GenBank mRNA | AF218416.1 | - | 4 - 5 | 661 | 105 |
AK004882.1 | - | 4 - 5 | 654 | 105 | |
AK050374.1 | - | 4 - 5 | 674 | 105 | |
BC026882.1 | - | 4 - 5 | 560 | 105 | |
BC099868.1 | - | 4 - 5 | 672 | 105 | |
BC099869.1 | - | 4 - 5 | 674 | 105 | |
BC099870.1 | - | 4 - 5 | 674 | 105 | |
BC099871.1 | - | 4 - 5 | 673 | 105 |