Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myo16
Entrez Gene ID
244281
Gene Name
myosin XVI
Gene Aliases
BM140241, C230040D10Rik, Gm1105, NYAP3, mKIAA0865
Chromosome Location
Chr.8: 10153916 - 10634742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081397.1 NP_001074866.1 25 - 26 3328 96
GenBank mRNA AB429291.1 - 22 - 23 2834 96
AK034257.1 - 2 - 3 202 96
AK220356.1 - 6 - 7 760 96
BC082311.1 - 25 - 26 3194 96
BC151049.1 - 25 - 26 3282 96
BC151051.1 - 25 - 26 3283 96
BC157966.1 - 25 - 26 3283 96
BC172122.1 - 25 - 26 3283 96

View Full Product Details