Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myo16
Entrez Gene ID
244281
Gene Name
myosin XVI
Gene Aliases
BM140241, C230040D10Rik, Gm1105, NYAP3, mKIAA0865
Chromosome Location
Chr.8: 10153916 - 10634742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081397.1 NP_001074866.1 2 - 3 586 107
GenBank mRNA AB429291.1 - 1 - 2 260 107
AK082350.1 - 2 - 3 322 107
BC072580.1 - 2 - 3 598 107
BC082311.1 - 2 - 3 454 107
BC151049.1 - 2 - 3 540 107
BC151051.1 - 2 - 3 541 107
BC157966.1 - 2 - 3 541 107
BC172122.1 - 2 - 3 541 107

View Full Product Details