The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007834.3 | NP_031860.1 | 1 - 2 | 291 | 81 |
GenBank mRNA | AB001990.1 | - | 1 - 2 | 106 | 81 |
AK077485.1 | - | 1 - 2 | 105 | 81 | |
AK077628.1 | - | 1 - 2 | 97 | 81 | |
BC003740.1 | - | 1 - 2 | 79 | 81 | |
BU709449.1 | - | 1 - 2 | 493 | 81 |