Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crmp1
Entrez Gene ID
12933
Gene Name
collapsin response mediator protein 1
Gene Aliases
CRMP-1, DRP-1, Dpysl1, ULIP-3, Ulip3
Chromosome Location
Chr.5: 37242058 - 37292163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136058.2 NP_001129530.1 2 - 3 547 75
NM_007765.4 NP_031791.3 2 - 3 412 75
GenBank mRNA AB006714.1 - 2 - 3 384 75
AF501323.1 - 2 - 3 665 75
AK158793.1 - 2 - 3 491 75
AK159049.1 - 2 - 3 356 75
BC031738.1 - 2 - 3 341 75
BC065046.1 - 2 - 3 525 75
CA321187.1 - 2 - 3 547 75
CX562443.1 - 2 - 3 413 75
U72875.1 - 2 - 3 157 75
Y09080.1 - 2 - 3 387 75

View Full Product Details