Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf32
Entrez Gene ID
56874
Gene Name
ring finger protein 32
Gene Aliases
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2
Chromosome Location
Chr.5: 29195170 - 29225526 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289757.1 NP_001276686.1 4 - 5 551 92
NM_021470.6 NP_067445.3 4 - 5 523 92
GenBank mRNA AF190664.1 - 1 - 2 285 92
AK006379.1 - 4 - 5 92
AK016026.1 - 4 - 5 494 92
BC050964.1 - 4 - 5 466 92
BC089493.1 - 4 - 5 466 92

View Full Product Details