Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ociad1
Entrez Gene ID
68095
Gene Name
OCIA domain containing 1
Gene Aliases
6030432N09Rik, AI481327, AW557942, Asrij, B230209J16Rik, BB021357, Emi2, Imi2, OCIA, TPA018
Chromosome Location
Chr.5: 73292772 - 73314077 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159887.1 NP_001153359.1 7 - 8 739 94
GenBank mRNA AY208935.1 - 7 - 8 638 94
CX238085.1 - 7 - 8 687 94

View Full Product Details