The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026770.1 | NP_081046.1 | 1 - 2 | 121 | 56 |
GenBank mRNA | BC023116.1 | - | 1 - 2 | 121 | 56 |
BQ555687.1 | - | 1 - 2 | 237 | 56 |