Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtn3
Entrez Gene ID
20168
Gene Name
reticulon 3
Gene Aliases
RTN3-A1
Chromosome Location
Chr.19: 7425895 - 7483291 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003934.2 NP_001003934.1 1 - 2 355 154
NM_001271486.1 NP_001258415.1 1 - 2 355 154
NM_001271487.1 NP_001258416.1 1 - 2 355 154
GenBank mRNA AK032109.1 - 1 - 2 332 154
AY750849.1 - 1 - 2 336 154
BC079882.1 - 1 - 2 265 154
BI729783.1 - - 268 155
BY295849.1 - 1 - 2 355 154

View Full Product Details