Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dntt
Entrez Gene ID
21673
Gene Name
deoxynucleotidyltransferase, terminal
Gene Aliases
Tdt
Chromosome Location
Chr.19: 41029275 - 41059525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043228.1 NP_001036693.1 5 - 6 920 96
NM_009345.2 NP_033371.2 5 - 6 920 96
GenBank mRNA AF316014.1 - 5 - 6 773 96
AF316015.1 - 5 - 6 773 96
AK039804.1 - 5 - 6 811 96
AK087978.1 - 5 - 6 921 96
AK088709.1 - 5 - 6 921 96
AK134009.1 - 5 - 6 814 96
X04123.1 - 5 - 6 807 96
X68670.2 - 5 - 6 802 96

View Full Product Details