Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dntt
Entrez Gene ID
21673
Gene Name
deoxynucleotidyltransferase, terminal
Gene Aliases
Tdt
Chromosome Location
Chr.19: 41029275 - 41059525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043228.1 NP_001036693.1 8 - 9 1275 74
NM_009345.2 NP_033371.2 8 - 9 1275 74
GenBank mRNA AF316014.1 - 8 - 9 1128 74
AF316015.1 - 8 - 9 1128 74
AK087978.1 - 8 - 9 1276 74
AK088709.1 - 8 - 9 1276 74
AK134009.1 - 8 - 9 1169 74
X68670.2 - 8 - 9 1157 74

View Full Product Details