Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam3
Entrez Gene ID
11497
Gene Name
a disintegrin and metallopeptidase domain 3 (cyritestin)
Gene Aliases
Cyrn1, Taz83, tMDC
Chromosome Location
Chr.8: 24677232 - 24726044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009619.4 NP_033749.2 4 - 5 341 78
GenBank mRNA BC060975.1 - 4 - 5 304 78
BC129920.1 - 4 - 5 314 78
X64227.1 - 4 - 5 351 78

View Full Product Details