The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198859.1 | NP_001185788.1 | 2 - 3 | 380 | 81 |
NM_001198860.1 | NP_001185789.1 | 3 - 4 | 573 | 81 | |
NM_001198861.1 | NP_001185790.1 | 3 - 4 | 573 | 81 | |
NM_013502.3 | NP_038530.1 | 2 - 3 | 380 | 81 | |
GenBank mRNA | AB033122.1 | - | 2 - 3 | 345 | 81 |
AJ010483.1 | - | 2 - 3 | 293 | 81 | |
AK133816.1 | - | 3 - 4 | 544 | 81 | |
AK149906.1 | - | 3 - 4 | 551 | 81 | |
AK160658.1 | - | 3 - 4 | 563 | 81 | |
AK165276.1 | - | 3 - 4 | 528 | 81 | |
AK170133.1 | - | 1 - 2 | 87 | 81 | |
AK178530.1 | - | 1 - 2 | 89 | 81 | |
AK179724.1 | - | 1 - 2 | 116 | 81 | |
AK214870.1 | - | 1 - 2 | 89 | 81 | |
AK218288.1 | - | 1 - 2 | 92 | 81 | |
AK219771.1 | - | 1 - 2 | 89 | 81 | |
BC013702.1 | - | 2 - 3 | 293 | 81 | |
BC015071.1 | - | 2 - 3 | 349 | 81 | |
BC042425.1 | - | 3 - 4 | 554 | 81 |