Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gfm1
Entrez Gene ID
28030
Gene Name
G elongation factor, mitochondrial 1
Gene Aliases
AW545374, D3Wsu133e, Gfm
Chromosome Location
Chr.3: 67430115 - 67475068 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138591.2 NP_613057.2 14 - 15 1882 93
GenBank mRNA AF315511.1 - 14 - 15 1849 93
BC013093.1 - 14 - 15 1885 93
BC031772.1 - 14 - 15 1849 93

View Full Product Details