Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcrl5
Entrez Gene ID
329693
Gene Name
Fc receptor-like 5
Gene Aliases
Fcrh3, mBXMH2
Chromosome Location
Chr.3: 87435782 - 87500678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113238.1 NP_001106709.1 10 - 11 1572 65
NM_183222.3 NP_899045.3 11 - 12 1836 65
GenBank mRNA AK089756.1 - 10 - 11 1542 65
AY158090.1 - 11 - 12 1833 65
AY506558.1 - 11 - 12 1842 65
BC137997.1 - 11 - 12 1770 65
BC137998.1 - 11 - 12 1770 65

View Full Product Details