Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pan3
Entrez Gene ID
72587
Gene Name
PAN3 polyA specific ribonuclease subunit homolog (S. cerevisiae)
Gene Aliases
2700050F09Rik, A430027N15Rik, AU014670
Chromosome Location
Chr.5: 147430162 - 147548502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028291.4 NP_082567.3 13 - 14 1894 75
GenBank mRNA AK012409.1 - 2 - 3 290 75
BC061486.1 - 2 - 3 487 75
BC082547.1 - 12 - 13 1423 75
BC086321.1 - 6 - 7 631 75

View Full Product Details