Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kl
Entrez Gene ID
16591
Gene Name
klotho
Gene Aliases
alpha-kl
Chromosome Location
Chr.5: 150952607 - 150993817 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013823.2 NP_038851.2 3 - 4 1715 96
GenBank mRNA AB005141.1 - 3 - 4 1623 96
BC138258.1 - 3 - 4 1715 96
BC138259.1 - 3 - 4 1715 96

View Full Product Details