The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001492.2 | NP_001001492.2 | 2 - 3 | 227 | 90 |
GenBank mRNA | AK029554.1 | - | 2 - 3 | 221 | 90 |
AK133316.1 | - | 2 - 3 | 227 | 90 | |
BC141285.1 | - | 1 - 2 | 77 | 90 | |
BC145611.1 | - | 1 - 2 | 77 | 90 |