The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025624.2 | NP_079900.1 | 1 - 2 | 67 | 75 |
GenBank mRNA | AK003439.1 | - | 1 - 2 | 60 | 75 |
AK011115.1 | - | 1 - 2 | 68 | 75 | |
AK013934.1 | - | 1 - 2 | 61 | 75 | |
AK167958.1 | - | 1 - 2 | 68 | 75 | |
BC027016.1 | - | 1 - 2 | 58 | 75 |