Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubl3
Entrez Gene ID
24109
Gene Name
ubiquitin-like 3
Gene Aliases
AW108023, HCG
Chromosome Location
Chr.5: 148504631 - 148552788 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC024507.1 - 1 - 2 157 129
BC043729.1 - 1 - 2 157 129

View Full Product Details