Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubl3
Entrez Gene ID
24109
Gene Name
ubiquitin-like 3
Gene Aliases
AW108023, HCG
Chromosome Location
Chr.5: 148504631 - 148552788 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011908.2 NP_036038.1 4 - 5 1116 60
GenBank mRNA AF044222.1 - 4 - 5 1058 60
AK145952.1 - 4 - 5 795 60
AK197397.1 - 1 - 2 98 60
AK204771.1 - 1 - 2 98 60
BC024507.1 - 6 - 7 966 60
BC025595.1 - 4 - 5 771 60
BC043729.1 - 6 - 7 966 60

View Full Product Details