Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slmap
Entrez Gene ID
83997
Gene Name
sarcolemma associated protein
Gene Aliases
D330001L02Rik, Slap, mKIAA1601
Chromosome Location
Chr.14: 26413168 - 26534624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032008.3 NP_114397.3 3 - 4 636 110
GenBank mRNA AK052158.1 - 4 - 5 1604 110
AK129403.1 - 3 - 4 636 110
AK141597.1 - 3 - 4 1793 110
AK164911.1 - 3 - 4 1791 110
AK166396.1 - 3 - 4 1792 110

View Full Product Details