Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slmap
Entrez Gene ID
83997
Gene Name
sarcolemma associated protein
Gene Aliases
D330001L02Rik, Slap, mKIAA1601
Chromosome Location
Chr.14: 26413168 - 26534624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032008.3 NP_114397.3 5 - 6 730 102
GenBank mRNA AK129403.1 - 5 - 6 730 102
AK141597.1 - 5 - 6 1887 102
AK164911.1 - 5 - 6 1885 102
AK166396.1 - 5 - 6 1886 102

View Full Product Details