The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081422.2 | NP_001074891.1 | 25 - 26 | 8982 | 63 |
GenBank mRNA | AF013969.1 | - | 16 - 17 | 4441 | 63 |